Order Entry
Netherlands
ContactUsLinkComponent
α,α,α-Trifluoro-4-iodotoluene ≥97.0% (by GC) stabilised
Irritant
α,α,α-Trifluoro-4-iodotoluene ≥97.0% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
Irritant
α,α,α-Trifluoro-4-iodotoluene ≥97.0% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together