Order Entry
Netherlands
ContactUsLinkComponent
α,α,α-Trifluoro-2-iodotoluene ≥98.0% (by GC) stabilised
Irritant
α,α,α-Trifluoro-2-iodotoluene ≥98.0% (by GC) stabilised
Catalog # TCIAI0383-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
α,α,α-Trifluoro-2-iodotoluene ≥98.0% (by GC) stabilised
Catalog # TCIAI0383-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:198°C
Density:1,896 g/cm³ (26 °C)
Flash Pt:81°C
 :Stabilised with copper chip
 :272.01 g/mol
EINECS:207-147-1
 :GHS07
CAS Number:444-29-1
Formula:C₇H₄F₃I
MDL Number:MFCD00001041