

2,3,4,5,6-Pentafluorotoluene ≥98.0% (by GC)
: TCI
:771-56-2
:
:C₇H₃F₅
:MFCD00000298
:182.09 g/mol
:116 - 117 °C (1013 hPa)
:1,439 g/cm³ (20 °C)
:34°C
:Refrigerator
:212-233-7
:1993
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together