
2,6-Difluorotoluene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:443-84-5
:
Formula:C₇H₆F₂
MDL Number:MFCD00043898
Boiling Pt:112°C
Density:1,1296 g/cm³ (20 °C)
Flash Pt:10°C
UN:1993
ADR:3,II
:128.12 g/mol
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Related Information
Frequently Bought Together