
4-Chloro-2-methylanisole ≥97.0% (by GC)
Catalog # TCIAC1724-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:35.0 to 39.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:156.61 g/mol
Boiling Pt:212 - 214 °C (1013 hPa)
Melting Pt:37 °C
EINECS:221-860-5
:GHS07
CAS Number:3260-85-3
Formula:C₈H₉ClO
MDL Number:MFCD00070771