


4-Fluorobenzyl chloride ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:352-11-4
:
Formula:C₇H₆ClF
MDL Number:MFCD00000913
:144.58 g/mol
Boiling Pt:185°C
Melting Pt:–18 °C
Density:1,21 g/cm³ (20 °C)
Flash Pt:60°C
Storage Temperature:Ambient
EINECS:206-516-4
UN:2920
ADR:8,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together