Order Entry
Netherlands
ContactUsLinkComponent
2,4-Dichlorobenzotrifluoride ≥98.0% (by GC)
Corrosive
2,4-Dichlorobenzotrifluoride ≥98.0% (by GC)
Catalog # TCIAD1316-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
2,4-Dichlorobenzotrifluoride ≥98.0% (by GC)
Catalog # TCIAD1316-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :215 g/mol
Boiling Pt:118°C
Melting Pt:–1 °C
Density:1,4929 g/cm³ (20 °C)
Flash Pt:72°C
EINECS:206-278-1
UN:1760
ADR:8,III
 :GHS05
CAS Number:320-60-5
Formula:C₇H₃Cl₂F₃
MDL Number:MFCD00000582