4-Chlorotoluene ≥98.0% (by GC)
: TCI
:106-43-4
:
:C₇H₇Cl
:MFCD00000631
:126.59 g/mol
:14,02171
:162°C
:6…8 °C
:1,07 g/cm³ (20 °C)
:53°C
:203-397-0
:2238
:3,III
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Appearance:White or Colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together