Order Entry
Netherlands
ContactUsLinkComponent
m-Toluic acid ≥98.0% (by titrimetric analysis)
Irritant
m-Toluic acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
m-Toluic acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    110.0 to 113.0 deg-C

Specification Test Results

Frequently Bought Together