
2-Bromo-5-(trifluoromethyl)aniline ≥97.0% (by GC)
TCIAA1483-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
:
:96°C
:47…49 °C
:1,694 g/cm³ (25 °C)
:6.1,III
:240.02 g/mol
:207-227-6
:GHS07
:454-79-5
:C₇H₅BrF₃N
:MFCD00007637