
2-Bromo-4-(trifluoromethyl)aniline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:57946-63-1
:
Formula:C₇H₅BrF₃N
MDL Number:MFCD00042150
Boiling Pt:109 - 110 °C (10 -rr)
Melting Pt:26…28 °C
Density:1,7 g/cm³ (20 °C)
EINECS:261-035-7
ADR:6.1,III
:240.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to lump
- Purity (GC):min. 98.0 %
- Melting point:32.0 to 36.0 deg-C
Specification Test Results
Frequently Bought Together