Order Entry
Netherlands
ContactUsLinkComponent
5-Bromo-2-fluorotoluene ≥97.0% (by GC)
Irritant
5-Bromo-2-fluorotoluene ≥97.0% (by GC)
Catalog # TCIAB1679-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
5-Bromo-2-fluorotoluene ≥97.0% (by GC)
Catalog # TCIAB1679-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :189.03 g/mol
Boiling Pt:94 - 95 °C (66,7 hPa)
Density:1,48 g/cm³ (20 °C)
Flash Pt:73°C
EINECS:257-202-9
 :GHS07
Storage Temperature:Ambient
CAS Number:51437-00-4
Formula:C₇H₆BrF
MDL Number:MFCD00000343