Tetraphenylethylene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:632-51-9
:
Formula:C₂₆H₂₀
MDL Number:MFCD00004764
:332.44 g/mol
Boiling Pt:420 °C (1013 hPa)
Melting Pt:204…205 °C
Density:1,148 g/cm³ (20 °C)
EINECS:211-179-1
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:224.0 to 227.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together