2-(2,4-Difluorophenyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:391604-55-0
:
Formula:C₁₁H₇F₂N
MDL Number:MFCD07787526
:191.18 g/mol
Specification Test Results
- Appearance:White or Colorless to Light yellow to Light orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together