


p-Anisidinium chloride ≥99.0% (by HPLC)
TCIAA0490-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Water:within almost transparency
:
:159.62 g/mol
:243-657-0
:2811
:6.1,III
:GHS06
:GHS08
:GHS09
:20265-97-8
:C₇H₁₀ClNO
:MFCD00036388