
trans-2,3-Dibromo-2-butene-1,4-diol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:21285-46-1
:
Formula:C₄H₆Br₂O₂
MDL Number:MFCD00004698
:245.9 g/mol
EINECS:244-313-2
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 99.0 %
- Melting point:113.0 to 117.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together