


1-Naphthylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
TCIAN0056-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
:
:179.65 g/mol
:14,06398
:209-014-3
:2811
:6.1,III
:GHS06
:GHS08
:GHS09
:552-46-5
:C₁₀H₁₀ClN
:MFCD00036370