
Tryptamine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT0890-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:115.0 to 118.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:160.22 g/mol
Merck Index:14,09796
EINECS:200-510-5
:GHS07
Storage Temperature:Refrigerator
CAS Number:61-54-1
Formula:C₁₀H₁₂N₂
MDL Number:MFCD00005661