Dibutyl sulphoxide ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2168-93-6
:
Formula:C₈H₁₈OS
MDL Number:MFCD00002093
:162.3 g/mol
EINECS:218-511-4
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together