


2,4-Dinitroaniline ≥99.0% (by GC)
: TCI
:97-02-9
:
:C₆H₅N₃O₄
:MFCD00007151
:183.12 g/mol
:14,03270
:401 °C (1013 hPa)
:177…178 °C
:1,6 g/cm³ (20 °C)
:224 °C
:Ambient
:202-553-5
:1596
:6.1,II
:GHS06
:GHS08
:GHS09
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:178.0 to 182.0 deg-C
- Solubility in hot Methanol:almost transparency
Frequently Bought Together