


2,4-Dinitroaniline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:97-02-9
:
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007151
:183.12 g/mol
Merck Index:14,03270
Boiling Pt:401 °C (1013 hPa)
Melting Pt:177…178 °C
Density:1,6 g/cm³ (20 °C)
Flash Pt:224 °C
Storage Temperature:Ambient
EINECS:202-553-5
UN:1596
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:179.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together