Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,4,6-Triiodophenol ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
2,4,6-Triiodophenol ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    159.0 to 162.0 deg-C

 

Frequently Bought Together