3,5-Diiodosalicylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:133-91-5
:
Formula:C₇H₄I₂O₃
MDL Number:MFCD00002444
:389.92 g/mol
Merck Index:14,03190
Boiling Pt:381 °C (1013 hPa)
Melting Pt:230°C
Storage Temperature:Ambient
EINECS:205-124-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together