Order Entry
Netherlands
ContactUsLinkComponent
 
Toxic
4-Iodobenzonitrile ≥98.0% (by GC)
Catalog # TCIAI0661-5G
Supplier:  TCI
CAS Number:  
undefined
Toxic
4-Iodobenzonitrile ≥98.0% (by GC)
Catalog # TCIAI0661-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    125.0 to 129.0 deg-C

Specification Test Results

About this item

 :
 :229.02 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:125…128 °C
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:3058-39-7
Formula:C₇H₄IN
MDL Number:MFCD00051310