

2,5-Difluorophenol ≥95.0% (by GC)
Catalog # TCIAD2036-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 95.0 %
- Melting point:40.0 to 44.0 deg-C
Specification Test Results
About this item
:
:130.09 g/mol
Boiling Pt:145°C
Melting Pt:42 °C
UN:1325
ADR:4.1,III
:GHS02
:GHS07
CAS Number:2713-31-7
Formula:C₆H₄F₂O
MDL Number:MFCD00042501