
3-(Trifluoromethoxy)benzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1014-81-9
:
Formula:C₈H₅F₃O₃
MDL Number:MFCD00041501
:206.12 g/mol
Boiling Pt:272 °C (1013 hPa)
Melting Pt:91…92 °C
EINECS:213-802-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:90.0 to 94.0 deg-C
Specification Test Results
Frequently Bought Together