Order Entry
Netherlands
ContactUsLinkComponent
 
 
4-Fluorobenzoic acid ≥98.0% (by GC, titration analysis)
  TCIAF0112-25G
 :  TCI
 :  
undefined
 
4-Fluorobenzoic acid ≥98.0% (by GC, titration analysis)
  TCIAF0112-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    183.0 to 187.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :140.11 g/mol
 :14,04171
 :254 °C (1013 hPa)
 :185 °C
 :1,49 g/cm³ (20 °C)
 :≥ 100 °C
 :Ambient
 :207-259-0
 :GHS07
 :456-22-4
 :C₇H₅FO₂
 :MFCD00002530