
2-Fluorobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:445-29-4
:
Formula:C₇H₅FO₂
MDL Number:MFCD00002405
:140.11 g/mol
Boiling Pt:245 °C (1013 hPa)
Melting Pt:121…124 °C
Density:1,46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-158-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
Specification Test Results
Frequently Bought Together