
2,5-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1699-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Purple powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:130.0 to 132.0 deg-C
Specification Test Results
About this item
:
:158.1 g/mol
Boiling Pt:242 °C (1013 hPa)
Melting Pt:131…132 °C
EINECS:221-060-6
:GHS07
CAS Number:2991-28-8
Formula:C₇H₄F₂O₂
MDL Number:MFCD00002410