Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,4,6-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
  TCIAT2409-1G
 :  TCI
 :  
undefined
 
2,4,6-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
  TCIAT2409-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    142.0 to 146.0 deg-C

 

 

 :
 :176.09 g/mol
 :237 °C (1013 hPa)
 :144…145 °C
 :GHS07
 :28314-80-9
 :C₇H₃F₃O₂
 :MFCD00042398