
2,3,4-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:61079-72-9
:
Formula:C₇H₃F₃O₂
MDL Number:MFCD00061232
Boiling Pt:237 °C (1013 hPa)
Melting Pt:140…142 °C
:176.09 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:140.0 to 143.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together