Order Entry
Netherlands
ContactUsLinkComponent
2,3-Difluorobenzonitrile ≥97.0% (by GC)
Toxic
2,3-Difluorobenzonitrile ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Toxic
2,3-Difluorobenzonitrile ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together