


2,4,6-Trichlorophenol ≥97.0% (by GC)
: TCI
:88-06-2
:
:C₆H₃Cl₃O
:MFCD00002172
:197.45 g/mol
:14,09644
:120°C
:∼66 °C
:1,675 g/cm³ (25 °C)
:99 °C
:Ambient
:201-795-9
:2020
:6.1,III
:GHS07
:GHS08
:GHS09
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:65.0 to 69.0 deg-C
Frequently Bought Together