


Dicloran (2,6-dichloro-4-nitroaniline) ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:99-30-9
:
Formula:C₆H₄Cl₂N₂O₂
MDL Number:MFCD00007677
:207.02 g/mol
Boiling Pt:130°C
Melting Pt:186…189 °C
EINECS:202-746-4
ADR:6.1,III
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:190.0 to 195.0 deg-C
Specification Test Results
Frequently Bought Together