

2,4,5-Trichloroaniline ≥98.0% (by GC)
TCIAT0374-25G
: TCI
:
- Verpakking:Glass bottle
- VPE:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
- Solubility in Methanol:almost transparency
:
:196.46 g/mol
:262°C
:93…96 °C
:Kamertemperatuur
:211-254-9
:2811
:6.1,III
:GHS06
:GHS09
:636-30-6
:C₆H₄Cl₃N
:MFCD00007662