
1-Chloro-4-fluoro-2-nitrobenzene ≥98.0% (by GC)
TCIAC2166-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light yellow to Yellow to Orange powder to lump
- Purity (GC):min. 98.0 %
- Melting point:37.0 to 41.0 deg-C
:
:175.55 g/mol
:239°C
:37…40 °C
:206-456-9
:6.1,III
:GHS07
:345-17-5
:C₆H₃ClFNO₂
:MFCD00084853