
2-Chloro-6-fluorobenzoic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0992-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:155.0 to 159.0 deg-C
Specification Test Results
About this item
:
:174.56 g/mol
Boiling Pt:272 °C (1013 hPa)
Melting Pt:157…159 °C
EINECS:207-105-2
:GHS07
Storage Temperature:Ambient
CAS Number:434-75-3
Formula:C₇H₄ClFO₂
MDL Number:MFCD00002417