
2-Chloro-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
: TCI
:99-60-5
:
:C₇H₄ClNO₄
:MFCD00007209
:138…141 °C
:201.57 g/mol
:202-771-0
:GHS07
:Ambient
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:138.0 to 143.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together