
3,4-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)
TCIAD0340-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:205.0 to 209.0 deg-C
- Solubility in Methanol:almost transparency
:
:191.01 g/mol
:301 °C (1013 hPa)
:207…208 °C
:200-099-2
:GHS07
:Ambient
:51-44-5
:C₇H₄Cl₂O₂
:MFCD00002492