
2,3-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:50-45-3
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002413
Melting Pt:168.3 °C
:191.01 g/mol
EINECS:200-039-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:167.0 to 171.0 deg-C
Specification Test Results
Frequently Bought Together