
2,4-Dihydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0568-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:154.12 g/mol
Merck Index:14,08157
Boiling Pt:390 °C (1013 hPa)
Melting Pt:218…220 °C
Density:1,54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-946-9
:GHS07
CAS Number:89-86-1
Formula:C₇H₆O₄
MDL Number:MFCD00002451