
2,6-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
TCIAD1959-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
:
:182.18 g/mol
:315 °C (1013 hPa)
:180…182 °C
:1,365 g/cm³ (20 °C)
:Ambient
:215-985-4
:GHS07
:1466-76-8
:C₉H₁₀O₄
:MFCD00002437