
2,5-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2785-98-0
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002436
Boiling Pt:314 °C (1013 hPa)
Melting Pt:80 °C
:182.18 g/mol
EINECS:220-503-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:76.0 to 79.0 deg-C
Specification Test Results
Frequently Bought Together