4-(Methylamino)benzoic acid ≥98.0% (by HPLC, titration analysis)
: TCI
:10541-83-0
:
:C₈H₉NO₂
:MFCD00002535
:151.16 g/mol
:296 °C (1013 hPa)
:159 °C
:234-121-7
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:159.0 to 164.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together