
2,3,4-Trihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2144-08-3
:
Formula:C₇H₆O₄
MDL Number:MFCD00003325
:154.12 g/mol
Boiling Pt:332 °C (1013 hPa)
Melting Pt:159 °C
Density:1,054 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-404-2
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:160.0 to 164.0 deg-C
Specification Test Results
Frequently Bought Together