Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dimethoxybenzaldehyde ≥97.0% (by GC)
Irritant
2,6-Dimethoxybenzaldehyde ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
2,6-Dimethoxybenzaldehyde ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    96.0 to 99.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together