
2,5-Dimethoxybenzaldehyde ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:93-02-7
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00003314
:166.18 g/mol
Boiling Pt:270°C
Melting Pt:49…51 °C
Storage Temperature:Ambient
EINECS:202-211-5
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:48.0 to 52.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together