
2,4-Dibromophenol ≥98.0% (by GC, titration analysis)
: TCI
:615-58-7
:
:C₆H₄Br₂O
:MFCD00002149
:251.91 g/mol
:239°C
:38 °C
:2,07 g/cm³ (20 °C)
:210-436-5
:2811
:6.1,II
:GHS06
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:37.0 to 41.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together