

2-Bromo-4,6-dinitroaniline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1817-73-8
:
Formula:C₆H₄BrN₃O₄
MDL Number:MFCD00007146
:262.02 g/mol
Boiling Pt:401 °C (1013 hPa)
Melting Pt:154 °C
EINECS:217-329-2
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:151.0 to 154.0 deg-C
Specification Test Results
Frequently Bought Together