Order Entry
Netherlands
ContactUsLinkComponent
 
 
1-Bromo-4-fluoro-2-nitrobenzene ≥97.0% (by GC)
 :  TCI
 
undefined
 
1-Bromo-4-fluoro-2-nitrobenzene ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    39.0 to 43.0 deg-C

 

Frequently Bought Together